Search
Search

1700001K19Rik| Gene Symbol: 1700001K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025488.2 | NP_079764.2 | 4 - 5 | 319 | 78 | |
| GenBank mRNA | AK006098.1 | - | 4 - 5 | 320 | 78 | |
| AK006358.1 | - | 4 - 5 | 313 | 78 | ||
| BC048462.1 | - | 4 - 5 | 300 | 6774571 | 78 | |
Set Membership: |
|