Search
Search

Spryd7| Gene Symbol: Spryd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025697.3 | NP_079973.1 | 1 - 2 | 369 | 81 | |
| GenBank mRNA | AK018044.1 | - | 1 - 2 | 247 | 81 | |
| AK018153.1 | - | 1 - 2 | 370 | 81 | ||
| AK018290.1 | - | 1 - 2 | 219 | 81 | ||
| AK137685.1 | - | 218 | 81 | |||
| AK150789.1 | - | 1 - 2 | 244 | 81 | ||
| AK167306.1 | - | 1 - 2 | 211 | 81 | ||
| AK169059.1 | - | 1 - 2 | 218 | 81 | ||
| BC037015.1 | - | 1 - 2 | 203 | 5344409 | 81 | |
Set Membership: |
|