Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04208863_m1
          See other AVPI1 GE Assays ›
          Gene Symbol:
          Avpi1
          Gene Name:
          arginine vasopressin-induced 1
          Gene Aliases:
          2310008N12Rik, AA410041, VIT32, esu, mVIT32
          Chromosome Location:
          Chr.19: 42123275 - 42128993 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04208863_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Avpi1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027106.4 NP_081382.1 2 - 3 403 100
          GenBank mRNA AK009243.1 - 2 - 3 404 100
          BC027646.1 - 2 - 3 437 1344413 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001521.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Avpi1

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          cell cycle
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119206 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119207 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119203 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119205 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119204 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119202 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2120254 NM_027106.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2120254 AK009243.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2120254 AK177091.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2120254 AY505175.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2120254 BC027646.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 462146 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 210683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692187 AK003884.3 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692187 AK048669.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692187 AK154192.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692187 AK164964.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692187 BC042570.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692187 NM_176833.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 640340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 100974 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180389 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2120254 NM_027106.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2120254 AK009243.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2120254 AK177091.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2120254 AY505175.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2120254 BC027646.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261087 NM_027106.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261087 AK009243.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261087 AY505175.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119211 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180389 - -
          Whole Mouse Genome G4122A A_51_P261087 NM_027106.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261087 AK009243.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261087 AY505175.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132362 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180389 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14245462 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14245446 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14245455 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14245454 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14245463 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14245449 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119206 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119204 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119205 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119203 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119207 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14151176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14151186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14475321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14225309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14245445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14245449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14363697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14219441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14151185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119187 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10467741 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467740 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467743 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467742 - -
          Affymetrix HT MG-430 PM Array Plate 1423122_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10467739 - -
          Mouse Genome 430 2.0 Array 1423122_PM_at - -
          Mouse Genome 430 2.0 Array 1423122_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69534_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69534_at - -
          Affymetrix Mouse Clariom S Array PSR1900013422.mm.1 NM_027106.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013422.mm.1 NM_027106.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013422.mm.1 TC1900001521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900013420.mm.1 NM_027106.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013420.mm.1 NM_027106.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013420.mm.1 TC1900001521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900013421.mm.1 NM_027106.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013421.mm.1 NM_027106.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013421.mm.1 TC1900001521.mm.1 MTA TC
          Mouse Expression Set 430 1423122_at - -
          Mouse Expression Set 430 1423122_PM_at - -
          Murine Genome U74v2 Set 96866_PM_at - -
          Murine Genome U74v2 Set 96866_at - -
          Mouse Genome 430A 2.0 Array 1423122_PM_at - -
          Mouse Genome 430A 2.0 Array 1423122_at - -
          Affymetrix HT Mouse Genome 430A Array 1423122_at - -
          Affymetrix HT Mouse Genome 430A Array 1423122_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 523824 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1850377 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001850377 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2755585 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2755585 - -
          mouse-6 v1.1 (Illumina) scl52477.5.1_20-S - -
          Illumina Sentrix mouse-6.v1 bead array 1850377 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52477.5.1_20-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1850377 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2755585 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2813186 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2755585 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2813186 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline