Search
Search

Ccdc89| Gene Symbol: Ccdc89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027298.1 | NP_081574.1 | 1 | 1151 | 108 | |
| GenBank mRNA | AK006105.1 | - | 1 | 651 | 108 | |
| BC060150.1 | - | 1 | 885 | 108 | ||
Set Membership: |
|