Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04209071_g1
          See other CXX1C GE Assays ›
          Gene Symbol:
          Cxx1c
          Gene Name:
          CAAX box 1C
          Gene Aliases:
          2900027G03Rik, Mar8.1, Mart8a
          Chromosome Location:
          Chr.X: 53607922 - 53609135 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04209071_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cxx1c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028375.3 NP_082651.1 1 - 2 502 112
          GenBank mRNA AK214233.1 - 1 - 2 159 112
          CK627323.1 - 1 - 2 419 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000479.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cxx1c

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133266 NM_028375.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133266 BX529499.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133266 BC003436.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163928 NM_028375.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163928 AK013603.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163928 BC031701.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163928 BY123946.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163928 CK627323.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980257 NM_028375.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980257 BY123946.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105704 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163928 NM_028375.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163928 AK013603.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163928 BC031701.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163928 BY123946.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163928 CK627323.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133266 NM_028375.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133266 BX529499.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133266 BC003436.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980257 NM_028375.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980257 BY123946.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32388024 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105704 - -
          Whole Mouse Genome G4122A A_51_P285756 NM_028375.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285756 AK013603.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285756 AK214233.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285756 BC031701.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285756 CK627323.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138138 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105704 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14693908 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14693934 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14693912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32388018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14922847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14693919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32388003 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10599656 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599657 - -
          Affymetrix Mouse Gene 1.0 ST Array 10604634 - -
          Affymetrix Mouse Gene 1.0 ST Array 10604639 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599655 - -
          Affymetrix Mouse Gene 1.0 ST Array 10604638 - -
          Affymetrix Mouse Gene 1.0 ST Array 10604635 - -
          Affymetrix HT MG-430 PM Array Plate 1450080_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10599654 - -
          Affymetrix Mouse Gene 1.1 ST Array 10604637 - -
          Affymetrix Mouse Gene 1.1 ST Array 10604633 - -
          Mouse Genome 430 2.0 Array 1450080_PM_at - -
          Mouse Genome 430 2.0 Array 1450080_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72865_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72865_at - -
          Affymetrix Mouse Clariom S Array PSR0X00003139.mm.1 NM_028375.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003139.mm.1 NM_028375.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003139.mm.1 TC0X00000479.mm.1 MTA TC
          Mouse Expression Set 430 1450080_PM_at - -
          Mouse Expression Set 430 1450080_at - -
          Murine Genome U74v2 Set 166567_PM_f_at - -
          Murine Genome U74v2 Set 166567_f_at - -
          Mouse Genome 430A 2.0 Array 1450080_at - -
          Mouse Genome 430A 2.0 Array 1450080_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450080_at - -
          Affymetrix HT Mouse Genome 430A Array 1450080_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5220131 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220131 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2649511 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2649511 - -
          mouse-6 v1.1 (Illumina) scl072865.1_125-S - -
          Illumina Sentrix mouse-6.v1 bead array 5220131 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl072865.1_125-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220131 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2943972 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2943972 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248650 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl072865.1_125-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline