Search
Search

Pcdhga7| Gene Symbol: Pcdhga7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033590.3 | NP_291068.2 | 1 - 2 | 2543 | 67 | |
| GenBank mRNA | AY013801.1 | - | 1 - 2 | 2435 | 67 | |
| BC077712.1 | - | 1 - 2 | 2543 | 6825399 | 67 | |
Set Membership: |
|