Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04210368_m1
          See other SH2D1B1 GE Assays ›
          Gene Symbol:
          Sh2d1b1
          Gene Name:
          SH2 domain containing 1B1
          Gene Aliases:
          EAT-2, EAT-2A, Eat2, Eat2a, Sh2d1b
          Chromosome Location:
          Chr.1: 170277324 - 170285662 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04210368_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sh2d1b1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012009.4 NP_036139.3 410 111
          GenBank mRNA AK089191.1 - 415 111
          BC116698.1 - 410 40105516 111
          BC117508.1 - 410 40105515 111
          BC145540.1 - 416 9054103 111
          BC171965.1 - 416 9054251 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001568.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sh2d1b1

          Gene Ontology Categories:

          Function(s) Process(es)

          leukocyte activation involved in immune response
          positive regulation of natural killer cell mediated immunity
          natural killer cell inhibitory signaling pathway
          positive regulation of signal transduction
          peptidyl-tyrosine phosphorylation
          negative regulation of interferon-gamma production
          positive regulation of innate immune response
          negative regulation of natural killer cell mediated cytotoxicity
          positive regulation of natural killer cell mediated cytotoxicity
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of peptidyl-tyrosine phosphorylation
          phosphotyrosine binding
          protein binding
          signaling adaptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P496540 BC145540.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P496540 DQ131180.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P496540 DQ132507.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P496540 DQ132508.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P496540 DQ132509.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P496540 BC145540.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P496540 DQ131180.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P496540 DQ132507.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P496540 DQ132508.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P496540 DQ132509.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 775916 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 156352 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 170404 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 3766 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105102 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P496540 BC145540.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P496540 DQ131180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P496540 DQ132507.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P496540 DQ132508.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P496540 DQ132509.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P496540 BC145540.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P496540 DQ131180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P496540 DQ132507.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P496540 DQ132508.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P496540 DQ132509.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P496540 BC145540.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P496540 DQ131180.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P496540 DQ132507.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P496540 DQ132508.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P496540 DQ132509.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P496540 BC145540.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P496540 DQ131180.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P496540 DQ132507.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P496540 DQ132508.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P496540 DQ132509.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P286350 AK089191.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20821471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20821493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20821486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20821476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20821477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20821464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20821473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20821466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20821468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20821490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20821479 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105102 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00248983 - -
          Whole Mouse Genome G4122A A_52_P286350 AK089191.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P496540 BC145540.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P496540 DQ131180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P496540 DQ132507.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P496540 DQ132508.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P496540 DQ132509.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P496540 BC145540.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P496540 DQ131180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P496540 DQ132507.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P496540 DQ132508.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P496540 DQ132509.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108535 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106362 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105102 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00791661 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00791662 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00791663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20821478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00791662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00330138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00037694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00417337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00692817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01068188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00335135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00031282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00262483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00335143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00613209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01038051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01038054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00335147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00330140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00335146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00611855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00335141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20821511 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351480 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351479 - -
          Affymetrix HT MG-430 PM Array Plate 1423024_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10351477 - -
          Mouse Genome 430 2.0 Array 1423024_at - -
          Mouse Genome 430 2.0 Array 1423024_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100012841.mm.1 NM_012009.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012841.mm.1 NM_012009.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012841.mm.1 TC0100001568.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012842.mm.1 NM_012009.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012842.mm.1 NM_012009.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012842.mm.1 TC0100001568.mm.1 MTA TC
          Mouse Expression Set 430 1423024_at - -
          Mouse Expression Set 430 1423024_PM_at - -
          Murine Genome U74v2 Set 101343_PM_at - -
          Murine Genome U74v2 Set 101343_at - -
          Mouse Genome 430A 2.0 Array 1423024_PM_at - -
          Mouse Genome 430A 2.0 Array 1423024_at - -
          Affymetrix HT Mouse Genome 430A Array 1423024_at - -
          Affymetrix HT Mouse Genome 430A Array 1423024_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6420603 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006420603 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2671090 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2671090 - -
          mouse-6 v1.1 (Illumina) scl17245.5.1_164-S - -
          Illumina Sentrix mouse-6.v1 bead array 6420603 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl17245.5.1_164-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6420603 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2978617 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2978617 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline