Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04210801_sH
          See other OLFR1317 GE Assays ›
          Gene Symbol:
          Olfr1317
          Gene Name:
          olfactory receptor 1317
          Gene Aliases:
          MOR245-4
          Chromosome Location:
          Chr.2: 112141947 - 112142897 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04210801_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1317
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146448.1 NP_666659.1 1 857 99

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1317

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P117450 NM_146448.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P117450 NM_146448.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P348154 NM_146448.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21527500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21527502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21527490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21527476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21527480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21527497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21527494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21527472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21527492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21527501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21527477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21527474 - -
          Whole Mouse Genome G4122A A_51_P348154 NM_146448.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102637 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108681 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117450 NM_146448.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01347081 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01714398 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01347077 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01865240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02280958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01714389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01774653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01612826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527467 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01612822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527472 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10474523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474525 - -
          Affymetrix Mouse Gene 1.1 ST Array 10474524 - -
          Affymetrix Mouse Gene 1.1 ST Array 10474522 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 894674 - -
          ABI Mouse Genome Survey Microarray 899224 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5360044 - -
          Illumina mouse-6 v1.0 expression beadchip 5360044 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005360044 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2640357 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2640357 - -
          mouse-6 v1.1 (Illumina) scl4833.1.1_323-S - -
          Illumina Sentrix mouse-6.v1 bead array 5360044 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4833.1.1_323-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5360044 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2640357 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2640357 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline