Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04211251_s1
          See other OLFR1195 GE Assays ›
          Gene Symbol:
          Olfr1195
          Gene Name:
          olfactory receptor 1195
          Gene Aliases:
          MOR230-4
          Chromosome Location:
          Chr.2: 88682804 - 88683730 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04211251_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1195
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146753.2 NP_666964.1 1 756 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1195

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440030 NM_146753.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440030 NM_146753.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440030 NM_146753.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440030 NM_146753.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P440030 NM_146753.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P440030 NM_146753.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440030 NM_146753.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440030 NM_146753.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01194208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01617070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02100852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01617063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01194192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01617071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01798347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01617072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01617066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410688 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484753 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484752 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 550457 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4610672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004610672 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2760733 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2760733 - -
          mouse-6 v1.1 (Illumina) scl4297.1.1_152-S - -
          Illumina Sentrix mouse-6.v1 bead array 4610672 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4297.1.1_152-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4610672 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2760733 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2760733 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline