Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04211259_s1
          See other OLFR608 GE Assays ›
          Gene Symbol:
          Olfr608
          Gene Name:
          olfactory receptor 608
          Gene Aliases:
          MOR26-3
          Chromosome Location:
          Chr.7: 103470041 - 103470991 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04211259_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr608
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146756.2 NP_666967.2 1 653 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr608

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P388164 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124145 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P388164 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P388164 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124145 - -
          Whole Mouse Genome G4122A A_52_P388164 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126691 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06044665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06569188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383463 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555794 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555793 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 884882 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6650114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006650114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1243468 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243468 - -
          mouse-6 v1.1 (Illumina) scl32268.1.355_283-S - -
          Illumina Sentrix mouse-6.v1 bead array 6650114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32268.1.355_283-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6650114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1243468 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243468 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline