Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04211379_s1
          See other OLFR272 GE Assays ›
          Gene Symbol:
          Olfr272
          Gene Name:
          olfactory receptor 272
          Gene Aliases:
          MOR262-7
          Chromosome Location:
          Chr.4: 52910833 - 52911792 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04211379_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr272
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146839.1 NP_667050.1 1 546 98
          GenBank mRNA BC128006.1 - 1 550 40135971 98

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr272

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2108461 NM_146839.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2108461 BC128006.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158391 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2108461 NM_146839.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2108461 BC128006.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199249 NM_146839.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199249 BC128006.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22920032 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158391 - -
          Whole Mouse Genome G4122A A_51_P199249 NM_146839.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199249 BC128006.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130611 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158391 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03279473 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03279497 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03621087 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03621090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22920008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03761351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03444167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03444176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03279486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03444162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03444172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03444153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03279474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03444164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03444163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03812633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03812634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03444175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04017602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03783230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03783229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03444155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03812635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22920061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03621087 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10512946 - -
          Affymetrix Mouse Gene 1.1 ST Array 10512945 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 739308 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2510377 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002510377 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2758092 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2758092 - -
          mouse-6 v1.1 (Illumina) scl2442.1.1_198-S - -
          Illumina Sentrix mouse-6.v1 bead array 2510377 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl2442.1.1_198-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2510377 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2758092 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2758092 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline