Search
Search

Olfr206| Gene Symbol: Olfr206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146991.1 | NP_667202.1 | 1 | 453 | 173 | |
| GenBank mRNA | BC138396.1 | - | 1 | 480 | 8861416 | 173 |
| BC138397.1 | - | 1 | 480 | 8861417 | 173 | |
Set Membership: |
|