Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04211998_s1
          See other SOWAHC GE Assays ›
          Gene Symbol:
          Sowahc
          Gene Name:
          sosondowah ankyrin repeat domain family member C
          Gene Aliases:
          4921515A04Rik, 9430099J10Rik, Ankrd57, C820004L04Rik
          Chromosome Location:
          Chr.10: 59221922 - 59226434 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04211998_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sowahc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172939.3 NP_766527.3 1 3449 73
          GenBank mRNA AK050497.1 - 1 3420 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sowahc

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P98625 AK050497.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P98625 NM_172939.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P98625 AK050497.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P98625 NM_172939.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701179 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445537 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158345 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P98625 AK050497.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P98625 NM_172939.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P98625 AK050497.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P98625 NM_172939.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158345 - -
          Whole Mouse Genome G4122A A_52_P98625 AK050497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98625 NM_172939.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98625 AK050497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98625 NM_172939.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P21837 AK030944.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P21837 AK050497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P21837 NM_172939.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112335 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27167659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08368656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08664340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08551143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08674294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08356059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08730828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27167632 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10363349 - -
          Affymetrix Mouse Gene 1.0 ST Array 10363348 - -
          Affymetrix Mouse Gene 1.0 ST Array 10363347 - -
          Affymetrix HT MG-430 PM Array Plate 1454736_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10363346 - -
          Mouse Genome 430 2.0 Array 1454736_at - -
          Mouse Genome 430 2.0 Array 1454736_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 268301_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 268301_at - -
          Mouse Expression Set 430 1454736_PM_at - -
          Mouse Expression Set 430 1454736_at - -
          Murine Genome U74v2 Set 104494_at - -
          Murine Genome U74v2 Set 104494_PM_at - -
          Mouse Genome 430A 2.0 Array 1454736_at - -
          Mouse Genome 430A 2.0 Array 1454736_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1454736_at - -
          Affymetrix HT Mouse Genome 430A Array 1454736_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 628321 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7100520 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007100520 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2659796 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2659796 - -
          mouse-6 v1.1 (Illumina) scl00268301.1_190-S - -
          Illumina Sentrix mouse-6.v1 bead array 7100520 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00268301.1_190-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7100520 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00268301.1_190-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0