Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04212034_m1
          See other SLC28A2 GE Assays ›
          Gene Symbol:
          Slc28a2
          Gene Name:
          solute carrier family 28 (sodium-coupled nucleoside transporter), member 2
          Gene Aliases:
          2010208B10Rik, B430217P18, BB152493, Cnt2
          Chromosome Location:
          Chr.2: 122426355 - 122461152 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04212034_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc28a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172980.2 NP_766568.1 6 - 7 1777 72
          GenBank mRNA AF079853.2 - 5 - 6 72
          AK080957.1 - 6 - 7 1778 72
          AK152776.1 - 5 - 6 712 72
          BC055376.1 - 5 - 6 671 30077988 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001849.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc28a2

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          transport
          purine nucleoside transmembrane transport
          protein localization to cell surface
          sodium ion transmembrane transport
          nucleoside transmembrane transport
          nucleoside binding
          nucleoside transmembrane transporter activity
          nucleoside:sodium symporter activity
          purine nucleoside transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21586511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21586512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21586514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21586516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21586517 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111975 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 800874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272536 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 83867 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 200754 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 709922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573052 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 758609 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 73818 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103092 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153506 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091191 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265444 NM_001085518.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265444 AK152776.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265444 BC055376.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265444 AF079853.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265444 AK080957.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21586349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21586351 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04673377 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04673373 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103092 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153506 - -
          Whole Mouse Genome G4122A A_51_P265444 NM_001085518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265444 AK152776.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265444 BC055376.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265444 AF079853.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265444 AK080957.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140702 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103092 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153506 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04673373 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01849905 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21586511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21586512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21586516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21586514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21586517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21586408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01395559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01324865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01604021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01861155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01918819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01791850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01607498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01791855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02007735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01849905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01269708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01849901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01369331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21586508 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10475488 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475499 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475500 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475509 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475492 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475495 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475489 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475513 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475496 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475490 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475501 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475494 - -
          Affymetrix Mouse Gene 1.1 ST Array 10475487 - -
          Affymetrix Mouse Gene 1.1 ST Array 10475502 - -
          Mouse Genome 430 2.0 Array 1450639_PM_at - -
          Mouse Genome 430 2.0 Array 1450639_at - -
          Affymetrix Mouse Clariom S Array PSR0200014380.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014380.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014380.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014393.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014393.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014393.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014379.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014379.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014379.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014385.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014385.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014385.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014387.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014387.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014387.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014389.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014389.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014389.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014395.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014395.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014395.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014396.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014396.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014396.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014384.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014384.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014384.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014381.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014381.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014381.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014386.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014386.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014386.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014388.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014388.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014388.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014382.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014382.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014382.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014392.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014392.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014392.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014383.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014383.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014383.mm.1 TC0200001849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014394.mm.1 NM_172980.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014394.mm.1 NM_172980.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014394.mm.1 TC0200001849.mm.1 MTA TC
          Mouse Expression Set 430 1450639_PM_at - -
          Mouse Expression Set 430 1450639_at - -
          Mouse Genome 430A 2.0 Array 1450639_PM_at - -
          Mouse Genome 430A 2.0 Array 1450639_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2970242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002970242 - -
          mouse-6 v1.1 (Illumina) scl0269346.15_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 2970242 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0269346.15_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2970242 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2843286 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2843286 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2666935 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline