Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04212161_s1
          See other PSMB11 GE Assays ›
          Gene Symbol:
          Psmb11
          Gene Name:
          proteasome (prosome, macropain) subunit, beta type, 11
          Gene Aliases:
          5830406J20Rik, beta5t
          Chromosome Location:
          Chr.14: 54625310 - 54629556 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04212161_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmb11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175204.4 NP_780413.1 1 3640 106
          GenBank mRNA AK162727.1 - 1 3662 106

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmb11

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          proteasomal ubiquitin-independent protein catabolic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          proteolysis involved in cellular protein catabolic process
          endopeptidase activity
          threonine-type endopeptidase activity
          peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982890 AK037368.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982890 AK162727.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982890 NM_175204.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904039 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203245 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129583 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982890 AK037368.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982890 AK162727.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982890 NM_175204.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29613346 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129583 - -
          Whole Mouse Genome G4122A A_52_P366790 AK037368.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366790 AK162727.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366790 NM_175204.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208132 AK037368.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208132 AK041393.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208132 AK162727.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208132 NM_175204.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208132 AK017902.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112090 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129583 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11535912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29613339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11720602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11535914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11535905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11535910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11792985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613340 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10415087 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415089 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415088 - -
          Affymetrix HT MG-430 PM Array Plate 1453150_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10415086 - -
          Mouse Genome 430 2.0 Array 1453150_at - -
          Mouse Genome 430 2.0 Array 1453150_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73902_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73902_at - -
          Mouse Expression Set 430 1453150_at - -
          Mouse Expression Set 430 1453150_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453150_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453150_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 424582 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 780170 - -
          Illumina mouse-6 v1.0 expression beadchip 780170 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780170 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2632853 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2632853 - -
          mouse-6 v1.1 (Illumina) scl46300.2_81-S - -
          Illumina Sentrix mouse-6.v1 bead array 780170 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46300.2_81-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780170 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2632853 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline