| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100006125.mm.1 |
NM_001085440.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006125.mm.1 |
NM_001085440.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006125.mm.1 |
NM_175491.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006125.mm.1 |
NM_175491.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006125.mm.1 |
TC1100000763.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100006127.mm.1 |
NM_175491.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006127.mm.1 |
NM_175491.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006127.mm.1 |
TC1100000763.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100006126.mm.1 |
NM_001085440.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006126.mm.1 |
NM_001085440.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006126.mm.1 |
NM_175491.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006126.mm.1 |
NM_175491.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100006126.mm.1 |
TC1100000763.mm.1 |
MTA TC |