Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04212666_m1
          See other ISPD GE Assays ›
          Gene Symbol:
          Ispd
          Gene Name:
          isoprenoid synthase domain containing
          Gene Aliases:
          4930579E17Rik, AV040780
          Chromosome Location:
          Chr.12: 36380585 - 36689503 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04212666_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ispd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289502.1 NP_001276431.1 3 - 4 952 84
          NM_001289503.1 NP_001276432.1 4 - 5 684 84
          NM_001289504.1 NP_001276433.1 4 - 5 1102 84
          NM_178629.6 NP_848744.2 4 - 5 1102 84
          GenBank mRNA AK016317.1 - 4 - 5 651 84
          AK039004.1 - 4 - 5 970 84
          AK043122.1 - 4 - 5 685 84
          AK045940.1 - 4 - 5 1104 84
          AK051322.1 - 3 - 4 647 84
          BC024061.1 - 3 - 4 384 84
          BC050993.1 - 4 - 5 940 84
          BC086665.1 - 4 - 5 1033 30612014 84
          BC096030.1 - 4 - 5 998 30610161 84
          BC118030.1 - 2 - 3 84
          BC118038.1 - 4 - 5 870 40093984 84
          DV044370.1 - 3 - 4 505 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000324.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ispd

          Panther Classification:

          Molecular Function -

          transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          axon guidance
          isoprenoid biosynthetic process
          protein O-linked mannosylation
          telomerase activity
          RNA helicase activity
          catalytic activity
          DNA primase activity
          sulfate adenylyltransferase activity
          RNA guanylyltransferase activity
          tRNA guanylyltransferase activity
          mannose-phosphate guanylyltransferase activity
          transferase activity
          nucleotidyltransferase activity
          DNA/RNA helicase activity
          CTP:2,3-di-O-geranylgeranyl-sn-glycero-1-phosphate cytidyltransferase activity
          phospholactate guanylyltransferase activity
          ATP:coenzyme F420 adenylyltransferase activity
          2-C-methyl-D-erythritol 4-phosphate cytidylyltransferase activity
          UDP-N-acetylgalactosamine diphosphorylase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200002425.mm.1 NM_001289504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200002425.mm.1 TC1200000324.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200002424.mm.1 NM_001289502.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200002424.mm.1 NM_001289503.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200002424.mm.1 NM_001289504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200002424.mm.1 NM_178629.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1200002424.mm.1 TC1200000324.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200002423.mm.1 NM_001289502.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200002423.mm.1 NM_001289503.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200002423.mm.1 NM_001289504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200002423.mm.1 NM_178629.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1200002423.mm.1 TC1200000324.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline