Search
Search

Tmem241| Gene Symbol: Tmem241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289666.1 | NP_001276595.1 | 7 - 8 | 517 | 79 | |
| NM_001289667.1 | NP_001276596.1 | 5 - 6 | 357 | 79 | ||
| NM_178801.5 | NP_848916.2 | 7 - 8 | 517 | 79 | ||
| NR_110356.1 | 7 - 8 | 517 | 79 | |||
| GenBank mRNA | AK028973.1 | - | 67 | 79 | ||
| AK031517.1 | - | 7 - 8 | 511 | 79 | ||
| AK038419.1 | - | 7 - 8 | 495 | 79 | ||
| AK038472.1 | - | 7 - 8 | 462 | 79 | ||
| AK051727.1 | - | 8 - 9 | 668 | 79 | ||
| AK144963.1 | - | 7 - 8 | 519 | 79 | ||
| BC094382.1 | - | 7 - 8 | 433 | 30605060 | 79 | |
| BC137621.1 | - | 7 - 8 | 447 | 8860640 | 79 | |
| BC144788.1 | - | 5 - 6 | 288 | 79 | ||
Set Membership: |
|