Search
Search

BC055111| Gene Symbol: BC055111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183182.3 | NP_899005.2 | 1 - 2 | 109 | 113 | |
| GenBank mRNA | BC055111.1 | - | 1 - 2 | 109 | 6706604 | 113 |
Set Membership: |
|