| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0Y00000710.mm.1 |
NM_201530.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0Y00000710.mm.1 |
TC0Y00000154.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0Y00001640.mm.1 |
TC0Y00000284.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0Y00001642.mm.1 |
TC0Y00000284.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0Y00001629.mm.1 |
TC0Y00000283.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0Y00001097.mm.1 |
NM_001037748.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0Y00001097.mm.1 |
TC0Y00000206.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0Y00001098.mm.1 |
NM_001037748.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0Y00001098.mm.1 |
TC0Y00000206.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0Y00001630.mm.1 |
TC0Y00000283.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0Y00001628.mm.1 |
TC0Y00000283.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0Y00000713.mm.1 |
NM_201530.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0Y00000713.mm.1 |
TC0Y00000154.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0Y00000712.mm.1 |
NM_201530.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0Y00000712.mm.1 |
TC0Y00000154.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0Y00001639.mm.1 |
TC0Y00000284.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0Y00001095.mm.1 |
NM_001037748.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0Y00001095.mm.1 |
TC0Y00000206.mm.1 |
MTA TC |