Search
Search

Olfr1349| Gene Symbol: Olfr1349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207136.1 | NP_997019.1 | 1 | 545 | 104 | |
| GenBank mRNA | BC147396.1 | - | 1 | 567 | 9056466 | 104 |
| BC147397.1 | - | 1 | 567 | 9056451 | 104 | |
Set Membership: |
|