Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04213144_g1
          See other RSPH3A GE Assays ›
          Gene Symbol:
          Rsph3a
          Gene Name:
          radial spoke 3A homolog (Chlamydomonas)
          Gene Aliases:
          1700012G05Rik, 4930524H12Rik, R74860, Rshl2, Rshl2a
          Chromosome Location:
          Chr.17: 7945614 - 7979556 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04213144_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rsph3a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025789.5 NP_080065.4 8 1841 139
          GenBank mRNA AK148230.1 - 8 1803 139

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rsph3a

          Panther Classification:

          Molecular Function -

          structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957490 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367563 AK161285.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367563 BC110284.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367563 NM_001083945.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367563 AK006083.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367563 AK016473.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367563 AK161285.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367563 BC110284.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367563 NM_001083945.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367563 AK006083.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367563 AK016473.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048129 AK148230.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048129 NM_025789.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048129 AK148230.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048129 NM_025789.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114143 BC110424.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367563 AK161285.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367563 BC110284.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367563 NM_001083945.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367563 AK006083.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367563 AK016473.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367563 AK161285.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367563 BC110284.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367563 NM_001083945.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367563 AK006083.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367563 AK016473.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31023400 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08707958 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154837 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03273817 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08707949 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08707983 - -
          Whole Mouse Genome G4122A A_52_P507277 AK161285.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P507277 NM_001083945.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P507277 AK006083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P507277 AK016473.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P507277 AK076872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367563 AK161285.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367563 BC110284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367563 NM_001083945.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367563 AK006083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367563 AK016473.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367563 AK161285.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367563 BC110284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367563 NM_001083945.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367563 AK006083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367563 AK016473.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P376979 AK161285.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P376979 NM_001083945.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P376979 AK006083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P376979 AK016473.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P376979 BC054439.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116951 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125628 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08707958 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154837 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957525 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10441611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447622 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447623 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447628 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441617 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441612 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447620 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441615 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447621 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441616 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447624 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447625 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447627 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441613 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447626 - -
          Affymetrix HT MG-430 PM Array Plate 1432579_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432580_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10441610 - -
          Affymetrix Mouse Gene 1.1 ST Array 10447619 - -
          Mouse Genome 430 2.0 Array 1423119_PM_at - -
          Mouse Genome 430 2.0 Array 1432579_PM_at - -
          Mouse Genome 430 2.0 Array 1432579_at - -
          Mouse Genome 430 2.0 Array 1423119_at - -
          Mouse Genome 430 2.0 Array 1432580_PM_at - -
          Mouse Genome 430 2.0 Array 1432580_at - -
          Affymetrix Mouse Clariom S Array PSR1700000604.mm.1 NM_025789.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000604.mm.1 NM_025789.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000604.mm.1 TC1700000078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000610.mm.1 NM_025789.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000610.mm.1 NM_025789.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000610.mm.1 TC1700000078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000605.mm.1 NM_025789.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000605.mm.1 NM_025789.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000605.mm.1 TC1700000078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000609.mm.1 NM_025789.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000609.mm.1 NM_025789.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000609.mm.1 TC1700000078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000613.mm.1 NM_025789.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000613.mm.1 NM_025789.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000613.mm.1 TC1700000078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000607.mm.1 NM_025789.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000607.mm.1 NM_025789.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000607.mm.1 TC1700000078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000606.mm.1 NM_025789.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000606.mm.1 NM_025789.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000606.mm.1 TC1700000078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000608.mm.1 NM_025789.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000608.mm.1 NM_025789.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000608.mm.1 TC1700000078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000612.mm.1 NM_025789.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000612.mm.1 NM_025789.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000612.mm.1 TC1700000078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000611.mm.1 NM_025789.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000611.mm.1 NM_025789.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000611.mm.1 TC1700000078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000614.mm.1 NM_025789.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000614.mm.1 NM_025789.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000614.mm.1 TC1700000078.mm.1 MTA TC
          Mouse Expression Set 430 1423119_at - -
          Mouse Expression Set 430 1423119_PM_at - -
          Mouse Expression Set 430 1432580_at - -
          Mouse Expression Set 430 1432580_PM_at - -
          Mouse Expression Set 430 1432579_PM_at - -
          Mouse Expression Set 430 1432579_at - -
          Murine Genome U74v2 Set 96263_PM_at - -
          Murine Genome U74v2 Set 96263_at - -
          Mouse Genome 430A 2.0 Array 1423119_PM_at - -
          Mouse Genome 430A 2.0 Array 1423119_at - -
          Affymetrix HT Mouse Genome 430B Array 1432580_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432580_at - -
          Affymetrix HT Mouse Genome 430B Array 1432579_at - -
          Affymetrix HT Mouse Genome 430B Array 1432579_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6110014 - -
          Illumina mouse-6 v1.0 expression beadchip 6110014 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006110014 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2670271 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2670271 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230303 - -
          mouse-6 v1.1 (Illumina) scl51156.10.1_24-S - -
          Illumina Sentrix mouse-6.v1 bead array 6110014 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51156.10.1_24-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6110014 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2670271 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2670271 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl51156.10.1_24-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline