Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04213424_s1
          See other KRT31 GE Assays ›
          Gene Symbol:
          Krt31
          Gene Name:
          keratin 31
          Gene Aliases:
          Ha1, Kha1, Krt1-1, MKHA-1
          Chromosome Location:
          Chr.11: 100046646 - 100050551 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04213424_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krt31
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010659.2 NP_034789.2 7 1252 84
          GenBank mRNA M27734.1 - 7 1252 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krt31

          Gene Ontology Categories:

          Process(es)

          structural molecule activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140042 NM_010659.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140042 M27734.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938428 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 919046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159412 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163547 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140042 NM_010659.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140042 M27734.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P304200 NM_010659.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P304200 M27734.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28065708 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163547 - -
          Whole Mouse Genome G4122A A_51_P304200 NM_010659.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304200 M27734.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124684 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163547 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09260663 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09431559 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09431575 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08984731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28065614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08984729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09504108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09006943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09544522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09291383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09716657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09256117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28065699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09539599 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10390984 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390982 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390980 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390981 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390983 - -
          Affymetrix HT MG-430 PM Array Plate 1421589_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10390979 - -
          Mouse Genome 430 2.0 Array 1421589_at - -
          Mouse Genome 430 2.0 Array 1421589_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16660_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16660_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100034402.mm.1 NM_010659.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034402.mm.1 NM_010659.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034402.mm.1 TC1100003751.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034400.mm.1 NM_010659.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034400.mm.1 NM_010659.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034400.mm.1 TC1100003751.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034401.mm.1 NM_010659.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034401.mm.1 NM_010659.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034401.mm.1 TC1100003751.mm.1 MTA TC
          Mouse Expression Set 430 1421589_PM_at - -
          Mouse Expression Set 430 1421589_at - -
          Murine Genome U74v2 Set 103558_PM_at - -
          Murine Genome U74v2 Set 103558_at - -
          Mouse Genome 430A 2.0 Array 1421589_PM_at - -
          Mouse Genome 430A 2.0 Array 1421589_at - -
          Affymetrix HT Mouse Genome 430A Array 1421589_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421589_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 705033 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6450142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450142 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2622236 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2622236 - -
          mouse-6 v1.1 (Illumina) scl39577.7.1_81-S - -
          Illumina Sentrix mouse-6.v1 bead array 6450142 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39577.7.1_81-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2622235 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2622235 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline