| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700021628.mm.1 |
TC0700002457.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700021628.mm.1 |
NM_027403.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021628.mm.1 |
NM_027403.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021629.mm.1 |
TC0700002457.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700021629.mm.1 |
NM_027403.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021629.mm.1 |
NM_027403.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021621.mm.1 |
TC0700002457.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700021621.mm.1 |
NM_027403.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021621.mm.1 |
NM_027403.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021624.mm.1 |
TC0700002457.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700021624.mm.1 |
NM_027403.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021624.mm.1 |
NM_027403.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021632.mm.1 |
TC0700002457.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700021632.mm.1 |
NM_027403.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021632.mm.1 |
NM_027403.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021630.mm.1 |
TC0700002457.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700021630.mm.1 |
NM_027403.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021630.mm.1 |
NM_027403.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021622.mm.1 |
TC0700002457.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700021622.mm.1 |
NM_027403.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021622.mm.1 |
NM_027403.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021626.mm.1 |
TC0700002457.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700021626.mm.1 |
NM_027403.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700021626.mm.1 |
NM_027403.4 |
Refseq |