Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04213574_s1
          See other SPRR2B GE Assays ›
          Gene Symbol:
          Sprr2b
          Gene Name:
          small proline-rich protein 2B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 92316705 - 92318085 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04213574_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sprr2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011469.3 NP_035599.2 350 102
          GenBank mRNA AY158987.1 - 350 102
          BC132518.1 - 336 40130795 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000823.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sprr2b

          Gene Ontology Categories:

          Function(s) Process(es)

          epidermis development
          keratinocyte differentiation
          keratinization
          response to estradiol
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069760 NM_011469.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069760 AY158987.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069760 BC132518.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14052 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141827 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069760 NM_011469.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069760 AY158987.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069760 BC132518.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22330844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22330842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22330839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22330824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22330845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22330831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22330833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22330834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22330872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22330873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22330874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22330837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22330838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22330862 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141827 - -
          Whole Mouse Genome G4122A A_51_P459894 NM_011469.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459894 AY158987.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459894 BC132518.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108533 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141827 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02533669 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02533651 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02533644 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02533664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22330924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02533669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02455881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02455870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02700766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02559411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02854702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02455861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02533673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02533664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02533672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02473870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03069871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02740284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02533652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22330831 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493852 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493853 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493863 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493861 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493854 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493862 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493860 - -
          Affymetrix Mouse Clariom S Array PSR0300006389.mm.1 NM_011469.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006389.mm.1 NM_011469.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006389.mm.1 TC0300000823.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006390.mm.1 NM_011469.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006390.mm.1 NM_011469.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006390.mm.1 TC0300000823.mm.1 MTA TC
          Murine Genome U74v2 Set 101761_f_at - -
          Murine Genome U74v2 Set 99701_PM_f_at - -
          Murine Genome U74v2 Set 101761_PM_f_at - -
          Murine Genome U74v2 Set 99701_f_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 60037 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000060037 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248780 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248780 - -
          mouse-6 v1.1 (Illumina) scl020756.2_52-S - -
          Illumina Sentrix mouse-6.v1 bead array 60037 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020756.2_52-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 60037 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248780 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248780 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline