Search
Search

Pcdhga1| Gene Symbol: Pcdhga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033584.1 | NP_291062.1 | 1 | 1444 | 69 | |
| GenBank mRNA | AK037148.1 | - | 1 | 1595 | 69 | |
| AY013792.1 | - | 1 | 1444 | 69 | ||
Set Membership: |
|