| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700022986.mm.1 |
NM_146082.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022986.mm.1 |
NM_146082.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022986.mm.1 |
TC1700002502.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700022985.mm.1 |
NM_146082.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022985.mm.1 |
NM_146082.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022985.mm.1 |
TC1700002502.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700022984.mm.1 |
NM_146082.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022984.mm.1 |
NM_146082.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022984.mm.1 |
TC1700002502.mm.1 |
MTA TC |