Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04213917_s1
          See other RBM12 GE Assays ›
          Gene Symbol:
          Rbm12
          Gene Name:
          RNA binding motif protein 12
          Gene Aliases:
          5730420G12Rik, 9430070C08Rik, AI852903, SWAN, mKIAA0765
          Chromosome Location:
          Chr.2: 156094882 - 156111965 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04213917_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbm12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029397.3 NP_083673.3 3 3182 63
          NM_170598.2 NP_733486.2 3 3150 63
          GenBank mRNA AF345334.1 - 2 2971 63
          AF393215.1 - 3 3178 63
          AF393216.1 - 3 3146 63
          AK017591.1 - 1 1149 63
          AK029180.1 - 1 2448 63
          AK035014.1 - 3 3140 63
          AK080772.1 - 3 3140 63
          AK166833.1 - 1 2667 63
          BC026891.1 - 1 2586 63
          BC027810.1 - 1 1029 63
          BC052473.1 - 3 3125 6833669 63

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbm12

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          nucleotide binding
          nucleic acid binding
          RNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769575 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769581 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769577 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769578 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769579 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769580 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769582 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769576 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076001 BC052473.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076001 BC026891.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076001 BC027810.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076001 AK085111.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076001 AK166833.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112144 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076001 BC052473.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076001 BC026891.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076001 BC027810.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076001 AK085111.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076001 AK166833.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P276652 BC052473.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P276652 BC026891.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P276652 BC027810.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P276652 AK166833.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P276652 AK080772.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769563 - -
          Whole Mouse Genome G4122A A_51_P276652 BC052473.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276652 BC026891.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276652 BC027810.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276652 AK166833.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276652 AK080772.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138108 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01392677 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01392681 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769580 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769582 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769577 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769578 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769576 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769581 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769579 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21769584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01392681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01392677 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10489000 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489001 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489002 - -
          Affymetrix HT MG-430 PM Array Plate 1416929_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456964_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1416928_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10488982 - -
          Mouse Genome 430 2.0 Array 1457934_at - -
          Mouse Genome 430 2.0 Array 1456964_PM_at - -
          Mouse Genome 430 2.0 Array 1416929_at - -
          Mouse Genome 430 2.0 Array 1416929_PM_at - -
          Mouse Genome 430 2.0 Array 1416928_at - -
          Mouse Genome 430 2.0 Array 1457934_PM_at - -
          Mouse Genome 430 2.0 Array 1416928_PM_at - -
          Mouse Genome 430 2.0 Array 1456964_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75710_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75710_at - -
          Affymetrix Mouse Clariom S Array PSR0200042483.mm.1 NM_029397.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042483.mm.1 NM_029397.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042483.mm.1 NM_170598.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042483.mm.1 NM_170598.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042483.mm.1 TC0200005511.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200042481.mm.1 NM_029397.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042481.mm.1 NM_029397.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042481.mm.1 NM_170598.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042481.mm.1 NM_170598.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042481.mm.1 TC0200005511.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200042482.mm.1 NM_029397.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042482.mm.1 NM_029397.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042482.mm.1 NM_170598.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042482.mm.1 NM_170598.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200042482.mm.1 TC0200005511.mm.1 MTA TC
          Mouse Expression Set 430 1416928_at - -
          Mouse Expression Set 430 1416929_PM_at - -
          Mouse Expression Set 430 1416929_at - -
          Mouse Expression Set 430 1416928_PM_at - -
          Mouse Expression Set 430 1456964_PM_at - -
          Mouse Expression Set 430 1456964_at - -
          Murine Genome U74v2 Set 97199_at - -
          Murine Genome U74v2 Set 97199_PM_at - -
          Murine Genome U74v2 Set 169934_PM_i_at - -
          Murine Genome U74v2 Set 169934_i_at - -
          Mouse Genome 430A 2.0 Array 1416929_at - -
          Mouse Genome 430A 2.0 Array 1416928_at - -
          Mouse Genome 430A 2.0 Array 1416929_PM_at - -
          Mouse Genome 430A 2.0 Array 1416928_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416929_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416929_at - -
          Affymetrix HT Mouse Genome 430A Array 1416928_at - -
          Affymetrix HT Mouse Genome 430A Array 1416928_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456964_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456964_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6040411 - -
          Illumina mouse-6 v1.0 expression beadchip 6040411 - -
          Illumina mouse-6 v1.0 expression beadchip 6660129 - -
          Illumina mouse-6 v1.0 expression beadchip 4570239 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006660129 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006040411 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004570239 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1243646 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221822 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2707710 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221822 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2707710 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243646 - -
          mouse-6 v1.1 (Illumina) scl075710.1_90-S - -
          mouse-6 v1.1 (Illumina) scl0075710.1_219-S - -
          mouse-6 v1.1 (Illumina) scl0003069.1_215-S - -
          Illumina Sentrix mouse-6.v1 bead array 6040411 - -
          Illumina Sentrix mouse-6.v1 bead array 4570239 - -
          Illumina Sentrix mouse-6.v1 bead array 6660129 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003069.1_215-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0075710.1_219-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl075710.1_90-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4570239 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6040411 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6660129 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221822 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2626300 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2707710 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3104139 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3104139 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2626300 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2707710 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221822 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0075710.1_219-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl075710.1_90-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0