Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04213975_s1
          See other HBQ1A GE Assays ›
          Gene Symbol:
          Hbq1a
          Gene Name:
          hemoglobin, theta 1A
          Gene Aliases:
          A230090E05Rik, Gm876, Hbq1, Hbqt2
          Chromosome Location:
          Chr.11: 32300066 - 32304090 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04213975_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hbq1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175000.2 NP_778165.1 3 556 106
          GenBank mRNA AK039049.1 - 3 557 106
          BC139349.1 - 3 530 8862371 106
          BC139351.1 - 3 530 8862373 106

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hbq1a

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185869 NM_175000.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185869 AK039049.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185869 BC139349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185869 BC139351.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185869 NM_175000.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185869 AK039049.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185869 BC139349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185869 BC139351.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570639 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139539 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185869 NM_175000.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185869 AK039049.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185869 BC139349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185869 BC139351.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185869 NM_175000.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185869 AK039049.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185869 BC139349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185869 BC139351.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27688037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27688013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27688032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27688030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27688027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27687973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27687985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27688031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27688017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27687982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27687971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27687977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27688036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27687994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27687983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27687979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27688011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27687993 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139539 - -
          Whole Mouse Genome G4122A A_51_P185869 NM_175000.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185869 AK039049.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185869 BC139349.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185869 BC139351.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185869 NM_175000.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185869 AK039049.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185869 BC139349.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185869 BC139351.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107952 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27687985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27687993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27687977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27687992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27687983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27688092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27687982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09686333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09597594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09385498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09017665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09017660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09113495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09113487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27688049 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10375063 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375064 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375062 - -
          Affymetrix Mouse Clariom S Array PSR1100002860.mm.1 NM_175000.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100002860.mm.1 NM_175000.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100002860.mm.1 TC1100000340.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 606800 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2900619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002900619 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2673980 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2673980 - -
          mouse-6 v1.1 (Illumina) scl41727.3.1_77-S - -
          Illumina Sentrix mouse-6.v1 bead array 2900619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41727.3.1_77-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2900619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2673980 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2673980 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl41727.3.1_77-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline