Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04214000_s1
          See other FAM187B GE Assays ›
          Gene Symbol:
          Fam187b
          Gene Name:
          family with sequence similarity 187, member B
          Gene Aliases:
          1700020B09Rik, Tmem162
          Chromosome Location:
          Chr.7: 30973804 - 30989726 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04214000_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam187b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001242647.1 NP_001229576.1 2 1025 95
          NM_175240.4 NP_780449.2 2 1053 95
          NR_038860.1 2 787 95
          GenBank mRNA AK028081.1 - 2 1054 95
          AK148492.1 - 2 788 95
          BC019737.1 - 2 767 95
          BC120864.1 - 2 1054 40129787 95
          BC125560.1 - 2 1054 40130075 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam187b

          Panther Classification:

          Molecular Function -

          immunoglobulin superfamily cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440092 AK148492.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440092 NM_175240.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440092 AK028081.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440092 AK148492.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440092 NM_175240.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P440092 AK028081.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 51429 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140579 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649219 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944595 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778392 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461162 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176209 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100806 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155796 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100461 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440092 AK148492.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440092 NM_175240.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440092 AK028081.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440092 AK148492.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440092 NM_175240.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P440092 AK028081.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034402 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06156143 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100461 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100806 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155796 - -
          Whole Mouse Genome G4122A A_51_P440092 AK148492.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440092 NM_175240.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440092 AK028081.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440092 AK148492.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440092 NM_175240.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P440092 AK028081.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113894 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100806 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155796 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06331971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06356749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06353396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06561870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06561880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06561878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06443610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034440 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552072 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552074 - -
          Affymetrix HT MG-430 PM Array Plate 1453057_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552071 - -
          Mouse Genome 430 2.0 Array 1453057_at - -
          Mouse Genome 430 2.0 Array 1453057_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76415_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76415_at - -
          Affymetrix Mouse Clariom S Array PSR0700005223.mm.1 TC0700000583.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005223.mm.1 NM_001242647.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005223.mm.1 NM_001242647.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005227.mm.1 TC0700000583.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005227.mm.1 NM_001242647.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005227.mm.1 NM_001242647.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005227.mm.1 NM_175240.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005227.mm.1 NM_175240.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005227.mm.1 NR_038860.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005227.mm.1 NR_038860.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005222.mm.1 TC0700000583.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005222.mm.1 NM_001242647.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005222.mm.1 NM_001242647.1 Refseq
          Mouse Expression Set 430 1453057_PM_at - -
          Mouse Expression Set 430 1453057_at - -
          Murine Genome U74v2 Set 165810_i_at - -
          Murine Genome U74v2 Set 166069_PM_i_at - -
          Murine Genome U74v2 Set 165810_PM_i_at - -
          Murine Genome U74v2 Set 166069_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1453057_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453057_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 310840 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6380520 - -
          Illumina mouse-6 v1.0 expression beadchip 6380520 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380520 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233860 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233860 - -
          mouse-6 v1.1 (Illumina) scl32802.4.1_27-S - -
          Illumina Sentrix mouse-6.v1 bead array 6380520 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32802.4.1_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380520 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1233860 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233860 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl32802.4.1_27-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline