| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P256955 |
NM_177585.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P256955 |
AK052136.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P256955 |
NM_177585.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P256955 |
AK052136.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
881678 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
966236 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
186834 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
456771 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
429019 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
584282 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
315243 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
413144 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
957275 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
582617 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
243737 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
890303 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
279113 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
785679 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
963367 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
156908 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
745930 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
805274 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
403665 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563784 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
26746 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
274282 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
659865 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
105915 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
678843 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
200532 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
700304 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
766107 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
183507 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
607639 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
702390 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
908294 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
125282 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
609125 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
238560 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
961959 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
250472 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
226172 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
695118 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
165019 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
883831 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
261612 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
788343 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
611865 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
141815 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
840650 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
349751 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
294772 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
352861 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
12254 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
667642 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770273 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
694835 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
119777 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696812 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
641643 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
341028 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
542469 |
BC048756.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
542469 |
BC056434.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
358230 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
874931 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
296142 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
187250 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
626891 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
249051 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
297221 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
296322 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
421670 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
202803 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
621497 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
774156 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
299424 |
AK033066.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
275975 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
786825 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
467641 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
779704 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
166150 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
61970 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
259635 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
766033 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
959605 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
685891 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
298552 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889224 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
411385 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
689624 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
845391 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
871904 |
BC115506.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
871904 |
BC115507.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
871904 |
NM_134207.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
61577 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
171405 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
507454 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
269502 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
295510 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
535939 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
899904 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776018 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
444474 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
871328 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
709031 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
490542 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
14076 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
12536 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
150049 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
84343 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
171171 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
75691 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
21807 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
149707 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
168277 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
33775 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
35803 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
160393 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
135594 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
88182 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
52899 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
45163 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P133889 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P169255 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P256955 |
NM_177585.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P256955 |
AK052136.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P256955 |
NM_177585.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P256955 |
AK052136.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212236 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212261 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212278 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212276 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212247 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212272 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212268 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212263 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212257 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212234 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212249 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212264 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212255 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212235 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212274 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212277 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212252 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212242 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212283 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212245 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212259 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212254 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212266 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212270 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212237 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212281 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212284 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212240 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212280 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22212232 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938520 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938138 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938205 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938178 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938235 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938307 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938503 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938412 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938390 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P133889 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938247 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938478 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938437 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P169255 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00664322 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938269 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938193 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938318 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00664356 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00664510 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938369 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04938349 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P256955 |
NM_177585.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P256955 |
AK052136.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P256955 |
NM_177585.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P256955 |
AK052136.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P126642 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04938247 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04938318 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P169255 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P133889 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04938412 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04938369 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04938520 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04938478 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P00664322 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02849573 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02849564 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02849562 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02849555 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02849563 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212210 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212218 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212237 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212245 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212206 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212242 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212259 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212207 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212225 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212252 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212234 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212205 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212215 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212229 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212212 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212235 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212227 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212220 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212221 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212222 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212255 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212241 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212249 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212263 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212247 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212232 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212208 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212223 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212257 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212226 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212261 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212216 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22212254 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212242 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212209 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212222 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02849573 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212254 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212257 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212212 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212240 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212245 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212255 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212259 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02863399 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212221 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212218 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212249 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02863397 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212223 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212241 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212220 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02646543 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212216 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212252 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212225 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212235 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03073024 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212229 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212261 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02849568 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212210 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02863402 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22212215 |
- |
- |