Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04214093_sH
          See other SLFN8 GE Assays ›
          Gene Symbol:
          Slfn8
          Gene Name:
          schlafen 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 83002158 - 83021994 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04214093_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slfn8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001167743.1 NP_001161215.1 4 1921 97
          NM_181545.4 NP_853523.2 6 2849 97
          GenBank mRNA AK150875.1 - 4 2171 97
          AY261798.1 - 6 2849 97
          BC152549.1 - 4 1921 97

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slfn8

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117656 BC024709.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117656 NM_001167743.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117656 NM_181545.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117656 AY261798.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P105585 BC152549.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P105585 NM_001167743.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P105585 NM_181545.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P105585 AY261798.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 251400 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300735 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931946 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728761 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 688701 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 350672 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77815 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51181 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 39919 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131170 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117656 BC024709.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117656 NM_001167743.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117656 NM_181545.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117656 AY261798.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P105585 BC152549.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P105585 NM_001167743.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P105585 NM_181545.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P105585 AY261798.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27969652 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02379049 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02379042 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131170 - -
          Whole Mouse Genome G4122A A_52_P543489 BC152549.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P543489 NM_001167743.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P543489 NM_181545.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P543489 AK150875.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P543489 AY261798.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105585 BC152549.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105585 NM_001167743.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105585 NM_181545.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105585 AY261798.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131170 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02379042 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09341966 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09341984 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09341975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27969632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09273994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09739529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09273998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09007621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09007614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09282238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27969650 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10389144 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389153 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389159 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389145 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389150 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389152 - -
          Affymetrix HT MG-430 PM Array Plate 1451655_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10389151 - -
          Affymetrix Mouse Gene 1.1 ST Array 10389143 - -
          Affymetrix Mouse Gene 1.1 ST Array 10389134 - -
          Mouse Genome 430 2.0 Array 1451655_PM_at - -
          Mouse Genome 430 2.0 Array 1451655_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 276950_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 276950_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100031007.mm.1 NM_001167743.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031007.mm.1 NM_001167743.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031007.mm.1 NM_181545.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031007.mm.1 NM_181545.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031007.mm.1 TC1100003382.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100031008.mm.1 NM_001167743.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031008.mm.1 NM_001167743.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031008.mm.1 NM_181545.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031008.mm.1 NM_181545.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031008.mm.1 TC1100003382.mm.1 MTA TC
          Mouse Expression Set 430 1451655_PM_at - -
          Mouse Expression Set 430 1451655_at - -
          Murine Genome U74v2 Set 136580_at - -
          Murine Genome U74v2 Set 136580_PM_at - -
          Mouse Genome 430A 2.0 Array 1451655_at - -
          Mouse Genome 430A 2.0 Array 1451655_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451655_at - -
          Affymetrix HT Mouse Genome 430A Array 1451655_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 438018 - -
          ABI Mouse Genome Survey Microarray 836900 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104540167 - -
          Illumina mouse-6 v1.0 expression beadchip 104540167 - -
          Illumina mouse-6 v1.0 expression beadchip 670072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2596730 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2609979 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2596730 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2609979 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246175 - -
          mouse-6 v1.1 (Illumina) scl39825.6.1_191-S - -
          mouse-6 v1.1 (Illumina) GI_31581543-S - -
          Illumina Sentrix mouse-6.v1 bead array 670072 - -
          Illumina Sentrix mouse-6.v1 bead array 104540167 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39825.6.1_191-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670072 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104540167 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2596730 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2596730 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline