Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04214180_s1
          See other ZFP108 GE Assays ›
          Gene Symbol:
          Zfp108
          Gene Name:
          zinc finger protein 108
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 24254794 - 24262445 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04214180_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp108
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_018791.2 NP_061261.2 4 643 87
          GenBank mRNA AF167315.1 - 4 614 87
          AK032355.1 - 4 644 87
          BC100339.1 - 4 588 30939834 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp108

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P199008 BC100339.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P199008 AK012570.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P199008 AK032355.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P199008 NM_018791.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P199008 BC100339.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P199008 AK012570.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P199008 AK032355.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P199008 NM_018791.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204688 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307596 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 585029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127575 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134986 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P199008 BC100339.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P199008 AK012570.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P199008 AK032355.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P199008 NM_018791.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P199008 BC100339.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P199008 AK012570.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P199008 AK032355.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P199008 NM_018791.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199008 BC100339.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199008 AK012570.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199008 AK032355.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199008 NM_018791.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199008 BC100339.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199008 AK012570.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199008 AK032355.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199008 NM_018791.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199012 BC100339.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199012 AF167315.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199012 AK012570.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199012 AK032355.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199012 NM_018791.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004016 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06140450 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134986 - -
          Whole Mouse Genome G4122A A_51_P199012 BC100339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199012 AF167315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199012 AK012570.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199012 AK032355.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199012 NM_018791.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199008 BC100339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199008 AK012570.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199008 AK032355.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199008 NM_018791.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199008 BC100339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199008 AK012570.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199008 AK032355.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P199008 NM_018791.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135547 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134986 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06382876 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06382874 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06382882 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06382881 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06382873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06577556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06584520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05943681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06203399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06584515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06618843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06577555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06584527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06382874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06618844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06584517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004000 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550863 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550862 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550861 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550864 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498796 - -
          Affymetrix HT MG-430 PM Array Plate 1421454_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10498795 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550860 - -
          Mouse Genome 430 2.0 Array 1421454_PM_at - -
          Mouse Genome 430 2.0 Array 1421454_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54678_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54678_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700002712.mm.1 TC0700000374.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002712.mm.1 NM_018791.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002712.mm.1 NM_018791.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002707.mm.1 TC0700000374.mm.1 MTA TC
          Mouse Expression Set 430 1421454_PM_at - -
          Mouse Expression Set 430 1421454_at - -
          Mouse Genome 430A 2.0 Array 1421454_at - -
          Mouse Genome 430A 2.0 Array 1421454_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421454_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421454_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 353117 - -
          ABI Mouse Genome Survey Microarray 748763 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6350632 - -
          Illumina mouse-6 v1.0 expression beadchip 106020019 - -
          Illumina mouse-6 v1.0 expression beadchip 6350632 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006350632 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1243011 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243011 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2466586 - -
          mouse-6 v1.1 (Illumina) scl0054678.2_330-S - -
          mouse-6 v1.1 (Illumina) scl32971.4_155-S - -
          Illumina Sentrix mouse-6.v1 bead array 106020019 - -
          Illumina Sentrix mouse-6.v1 bead array 6350632 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0054678.2_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6350632 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106020019 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1243011 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243011 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline