Search
Search

Raet1d| Gene Symbol: Raet1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020030.2 | NP_064414.1 | 249 | 99 | ||
| GenBank mRNA | AF257520.1 | - | 3 | 251 | 99 | |
| BC106190.1 | - | 249 | 30142928 | 99 | ||
Set Membership: |
|