| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700001822.mm.1 |
NM_001104562.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001822.mm.1 |
NM_001104562.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001822.mm.1 |
TC1700000236.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001825.mm.1 |
NM_001104562.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001825.mm.1 |
NM_001104562.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001825.mm.1 |
TC1700000236.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001826.mm.1 |
NM_001104562.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001826.mm.1 |
NM_001104562.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001826.mm.1 |
TC1700000236.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001823.mm.1 |
NM_001104562.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001823.mm.1 |
NM_001104562.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001823.mm.1 |
TC1700000236.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001824.mm.1 |
NM_001104562.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001824.mm.1 |
NM_001104562.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001824.mm.1 |
TC1700000236.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001821.mm.1 |
NM_001104562.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001821.mm.1 |
NM_001104562.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001821.mm.1 |
TC1700000236.mm.1 |
MTA TC |