Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04214669_u1
          See other CD99 GE Assays ›
          Gene Symbol:
          Cd99
          Gene Name:
          CD99 antigen
          Gene Aliases:
          1110061M03Rik, 2410026K10Rik, D4, Pilr-l, pilr-1
          Chromosome Location:
          Chr.4|NT_187055: 48 - 1475 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04214669_u1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd99
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025584.2 NP_079860.2 155 81
          GenBank mRNA AB122023.1 - 155 81
          AK004342.1 - 157 81
          AK044051.1 - 148 81
          AY262355.1 - 81
          AY262356.1 - 122 81
          BC019482.1 - 153 4218841 81

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK004342.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd99

          Gene Ontology Categories:

          Function(s) Process(es)

          myeloid dendritic cell activation
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136732 AK010597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136732 AK044051.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136732 AY262355.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136732 AY262356.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136732 BC019482.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136732 AK010597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136732 AK044051.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136732 AY262355.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136732 AY262356.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136732 BC019482.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P372046 AK002762.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P351763 AK010597.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P351763 AK044051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P351763 AK004342.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P351763 AY262356.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P351763 BC019482.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10608701 - -
          Affymetrix HT MG-430 PM Array Plate 1453556_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1430514_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1428850_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10608701 - -
          Mouse Genome 430 2.0 Array 1430514_PM_a_at - -
          Mouse Genome 430 2.0 Array 1430514_a_at - -
          Mouse Genome 430 2.0 Array 1453556_PM_x_at - -
          Mouse Genome 430 2.0 Array 1428850_PM_x_at - -
          Mouse Genome 430 2.0 Array 1428850_x_at - -
          Mouse Genome 430 2.0 Array 1453556_x_at - -
          Mouse Expression Set 430 1453556_x_at - -
          Mouse Expression Set 430 1453556_PM_x_at - -
          Mouse Expression Set 430 1430514_a_at - -
          Mouse Expression Set 430 1430514_PM_a_at - -
          Mouse Expression Set 430 1428850_x_at - -
          Mouse Expression Set 430 1428850_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1430514_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1453556_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1428850_x_at - -
          Mouse Genome 430A 2.0 Array 1428850_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1453556_x_at - -
          Mouse Genome 430A 2.0 Array 1430514_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1453556_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1453556_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1430514_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1430514_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428850_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1428850_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3990411 - -
          Illumina mouse-6 v1.0 expression beadchip 3990411 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003990411 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232849 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232849 - -
          mouse-6 v1.1 (Illumina) scl0066478.1_151-S - -
          Illumina Sentrix mouse-6.v1 bead array 3990411 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0066478.1_151-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3990411 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232849 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0066478.1_151-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-rpnmj:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0