Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04214740_u1
          See other AHSP GE Assays ›
          Gene Symbol:
          Ahsp
          Gene Name:
          alpha hemoglobin stabilizing protein
          Gene Aliases:
          EDRF, Eraf
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04214740_u1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ahsp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133245.1 NP_573508.1 247 99
          GenBank mRNA AF060220.1 - 231 99
          AF364516.1 - 230 99
          AK011084.1 - 247 99
          AK028172.1 - 247 99
          AK146724.1 - 241 99
          BC110627.2 - 225 40051901 99

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ahsp

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          hemoglobin metabolic process
          hemopoiesis
          erythrocyte differentiation
          protein stabilization
          hemoglobin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P534583 AF364516.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P534583 AK011084.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P534583 AK028172.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P534583 AK146724.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P534583 BC110627.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P534583 AF364516.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P534583 AK011084.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P534583 AK028172.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P534583 AK146724.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P534583 BC110627.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P534583 AF364516.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P534583 AK011084.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P534583 AK028172.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P534583 AK146724.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P534583 BC110627.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P534583 AF364516.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P534583 AK011084.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P534583 AK028172.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P534583 AK146724.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P534583 BC110627.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534583 AF364516.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534583 AK011084.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534583 AK028172.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534583 AK146724.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534583 BC110627.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534583 AF364516.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534583 AK011084.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534583 AK028172.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534583 AK146724.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534583 BC110627.2 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10608710 - -
          Affymetrix HT MG-430 PM Array Plate 1449077_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10608710 - -
          Mouse Genome 430 2.0 Array 1449077_PM_at - -
          Mouse Genome 430 2.0 Array 1449077_at - -
          Mouse Expression Set 430 1449077_PM_at - -
          Mouse Expression Set 430 1449077_at - -
          Mouse Genome 430A 2.0 Array 1449077_PM_at - -
          Mouse Genome 430A 2.0 Array 1449077_at - -
          Affymetrix HT Mouse Genome 430A Array 1449077_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449077_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 680198 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2350242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002350242 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2619200 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2619200 - -
          mouse-6 v1.1 (Illumina) scl00170812.1_267-S - -
          Illumina Sentrix mouse-6.v1 bead array 2350242 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00170812.1_267-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2350242 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2619200 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2619200 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0