Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04214876_sH
          See other TMEM230 GE Assays ›
          Gene Symbol:
          Tmem230
          Gene Name:
          transmembrane protein 230
          Gene Aliases:
          5730494N06Rik, AA407821
          Chromosome Location:
          Chr.2: 132239492 - 132248415 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04214876_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem230
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001141971.1 NP_001135443.1 4 850 100
          NM_027478.3 NP_081754.2 4 910 100
          GenBank mRNA AK017729.1 - 4 904 100
          AK170062.1 - 4 883 100
          BC033429.1 - 4 881 5035633 100
          BC080755.1 - 3 2274 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem230

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21639322 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21639325 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961094 BC080755.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961094 AK170062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961094 BC033429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961094 AK017729.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961094 NM_001141971.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192141 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623270 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336785 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970057 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175937 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961094 BC080755.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961094 AK170062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961094 BC033429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961094 AK017729.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961094 NM_001141971.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21639336 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175937 - -
          Whole Mouse Genome G4122A A_51_P390586 BC080755.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390586 AK170062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390586 BC033429.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390586 AK017729.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390586 NM_001141971.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107829 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175937 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01906058 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01906055 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01906069 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01453720 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01453718 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21639322 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21639325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21639305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01819295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01453718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01906062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01836785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01540445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01448871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01836776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21639303 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10487928 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487929 - -
          Affymetrix HT MG-430 PM Array Plate 1452773_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10487927 - -
          Mouse Genome 430 2.0 Array 1452773_PM_at - -
          Mouse Genome 430 2.0 Array 1452773_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70612_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70612_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200040099.mm.1 NM_001141971.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040099.mm.1 NM_001141971.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040099.mm.1 NM_027478.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040099.mm.1 NM_027478.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040099.mm.1 TC0200004670.mm.1 MTA TC
          Mouse Expression Set 430 1452773_at - -
          Mouse Expression Set 430 1452773_PM_at - -
          Murine Genome U74v2 Set 97916_PM_at - -
          Murine Genome U74v2 Set 97916_at - -
          Mouse Genome 430A 2.0 Array 1452773_PM_at - -
          Mouse Genome 430A 2.0 Array 1452773_at - -
          Affymetrix HT Mouse Genome 430A Array 1452773_at - -
          Affymetrix HT Mouse Genome 430A Array 1452773_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 751134 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4570181 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004570181 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2663516 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2663516 - -
          mouse-6 v1.1 (Illumina) scl070612.1_126-S - -
          Illumina Sentrix mouse-6.v1 bead array 4570181 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl070612.1_126-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4570181 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2663516 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2663516 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl070612.1_126-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline