Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04231240_s1
          See other HAMP GE Assays ›
          Gene Symbol:
          Hamp
          Gene Name:
          hepcidin antimicrobial peptide
          Gene Aliases:
          Hamp1, Hepc, Hepc1
          Chromosome Location:
          Chr.7: 30942368 - 30944017 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04231240_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hamp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032541.1 NP_115930.1 3 240 86
          GenBank mRNA AF297664.1 - 3 240 86
          AF503444.1 - 3 240 86
          BC021587.1 - 3 223 5097807 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hamp

          Gene Ontology Categories:

          Function(s) Process(es)

          myeloid cell homeostasis
          cellular iron ion homeostasis
          JAK-STAT cascade
          negative regulation of iron ion transmembrane transport
          defense response to bacterium
          positive regulation of macrophage activation
          negative regulation of bone resorption
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of inflammatory response
          defense response to fungus
          iron ion homeostasis
          positive regulation of cellular protein catabolic process
          hormone activity
          copper ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P669922 AF297664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P669922 AF503444.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P669922 BC021587.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P669922 NM_032541.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P669922 AF297664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P669922 AF503444.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P669922 BC021587.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P669922 NM_032541.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2530 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136489 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P669922 AF297664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P669922 AF503444.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P669922 BC021587.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P669922 NM_032541.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P669922 AF297664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P669922 AF503444.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P669922 BC021587.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P669922 NM_032541.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25034169 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136489 - -
          Whole Mouse Genome G4122A A_52_P669922 AF297664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P669922 AF503444.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P669922 BC021587.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P669922 NM_032541.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P669922 AF297664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P669922 AF503444.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P669922 BC021587.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P669922 NM_032541.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106105 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136489 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06062309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25034222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06224316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25034172 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10562171 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562170 - -
          Affymetrix HT MG-430 PM Array Plate 1419196_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1419197_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10562169 - -
          Mouse Genome 430 2.0 Array 1456289_PM_at - -
          Mouse Genome 430 2.0 Array 1419196_PM_at - -
          Mouse Genome 430 2.0 Array 1419197_PM_x_at - -
          Mouse Genome 430 2.0 Array 1419196_at - -
          Mouse Genome 430 2.0 Array 1419197_x_at - -
          Mouse Genome 430 2.0 Array 1456289_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 84506_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 84506_at - -
          Affymetrix Mouse Clariom S Array PSR0700024862.mm.1 TC0700002783.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024862.mm.1 NM_032541.2 Refseq
          Mouse Expression Set 430 1419196_at - -
          Mouse Expression Set 430 1456289_PM_at - -
          Mouse Expression Set 430 1419197_x_at - -
          Mouse Expression Set 430 1419196_PM_at - -
          Mouse Expression Set 430 1419197_PM_x_at - -
          Mouse Expression Set 430 1456289_at - -
          Murine Genome U74v2 Set 104588_PM_at - -
          Murine Genome U74v2 Set 104588_at - -
          Mouse Genome 430A 2.0 Array 1419197_x_at - -
          Mouse Genome 430A 2.0 Array 1456289_at - -
          Mouse Genome 430A 2.0 Array 1419197_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1419196_at - -
          Mouse Genome 430A 2.0 Array 1419196_PM_at - -
          Mouse Genome 430A 2.0 Array 1456289_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419197_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1419196_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419196_at - -
          Affymetrix HT Mouse Genome 430A Array 1419197_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 303659 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4920112 - -
          Illumina mouse-6 v1.0 expression beadchip 4920112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004920112 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236304 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236304 - -
          mouse-6 v1.1 (Illumina) scl31505.3.1_46-S - -
          Illumina Sentrix mouse-6.v1 bead array 4920112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31505.3.1_46-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4920112 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1236304 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236304 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline