Search
Search

LOC106740| Gene Symbol: LOC106740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027905.1 | 1 | 349 | 93 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PHD finger protein 10 | Phf10 |
1810055P05Rik, AV024533 |
Sense | Chr. 17 - 14944994 - 14961638 on NCBI Build GRCm38 | |
| Downstream | predicted gene 3417 | Gm3417 | 15.4kb | Sense | Chr. 17 - 14964189 - 14978812 on NCBI Build GRCm38 |
Set Membership: |
|