Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04242377_s1
          See other DLX1AS GE Assays ›
          Gene Symbol:
          Dlx1as
          Gene Name:
          distal-less homeobox 1, antisense
          Gene Aliases:
          Dix1as, Dlx1os
          Chromosome Location:
          Chr.2: 71530638 - 71537891 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04242377_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dlx1as
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002854.2 2 2551 89
          GenBank mRNA AK132348.1 - 2 2552 89

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic cranial skeleton morphogenesis
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010579 BY713853.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25399 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010579 BY713853.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10483625 - -
          Affymetrix HT MG-430 PM Array Plate 1419845_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10483624 - -
          Mouse Genome 430 2.0 Array 1419845_at - -
          Mouse Genome 430 2.0 Array 1419845_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200046688.mm.1 NR_002854.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200046688.mm.1 NR_002854.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200046688.mm.1 TC0200005498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200031902.mm.1 NR_002854.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031902.mm.1 NR_002854.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031902.mm.1 TC0200005498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200031901.mm.1 NR_002854.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031901.mm.1 NR_002854.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031901.mm.1 TC0200005498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200031903.mm.1 NR_002854.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031903.mm.1 NR_002854.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031903.mm.1 TC0200005498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200031900.mm.1 NR_002854.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031900.mm.1 NR_002854.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031900.mm.1 TC0200005498.mm.1 MTA TC
          Mouse Expression Set 430 1419845_at - -
          Mouse Expression Set 430 1419845_PM_at - -
          Murine Genome U74v2 Set 97117_PM_at - -
          Murine Genome U74v2 Set 97117_at - -
          Mouse Genome 430A 2.0 Array 1419845_at - -
          Mouse Genome 430A 2.0 Array 1419845_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419845_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419845_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 455477 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2924781 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2924781 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline