Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04242850_m1
          See other FUT7 GE Assays ›
          Gene Symbol:
          Fut7
          Gene Name:
          fucosyltransferase 7
          Gene Aliases:
          AI853193, FTVII, Fuc-TVII, FucT-VII
          Chromosome Location:
          Chr.2: 25422351 - 25426374 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04242850_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fut7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177366.1 NP_001170837.1 1 - 2 272 79
          GenBank mRNA BC105670.1 - 1 - 2 236 40046875 79
          GQ424192.1 - 1 - 2 587 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000416.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation
          leukocyte migration involved in immune response
          protein glycosylation
          alpha-1,6-mannosyltransferase activity
          alpha-1,2-mannosyltransferase activity
          mannosyltransferase activity
          alpha-1,3-mannosyltransferase activity
          alpha-1,3-galactosyltransferase activity
          UDP-glucose:glycoprotein glucosyltransferase activity
          glycolipid mannosyltransferase activity
          oligosaccharyl transferase activity
          dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
          acetylglucosaminyltransferase activity
          acetylgalactosaminyltransferase activity
          galactosyltransferase activity
          fucosyltransferase activity
          O antigen polymerase activity
          lipopolysaccharide-1,6-galactosyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups
          cellulose synthase activity
          9-phenanthrol UDP-glucuronosyltransferase activity
          1-phenanthrol glycosyltransferase activity
          9-phenanthrol glycosyltransferase activity
          1,2-dihydroxy-phenanthrene glycosyltransferase activity
          phenanthrol glycosyltransferase activity
          beta-1,4-mannosyltransferase activity
          alpha-1,2-galactosyltransferase activity
          dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
          lipopolysaccharide-1,5-galactosyltransferase activity
          dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
          dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
          inositol phosphoceramide synthase activity
          glucosyltransferase activity
          alpha-(1->3)-fucosyltransferase activity
          alpha-(1->6)-fucosyltransferase activity
          indole-3-butyrate beta-glucosyltransferase activity
          salicylic acid glucosyltransferase (ester-forming) activity
          salicylic acid glucosyltransferase (glucoside-forming) activity
          benzoic acid glucosyltransferase activity
          chondroitin hydrolase activity
          dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
          cytokinin 9-beta-glucosyltransferase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200002413.mm.1 NM_001177366.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002413.mm.1 NM_001177366.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002413.mm.1 NM_001177367.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002413.mm.1 NM_001177367.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002413.mm.1 NM_001289455.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002413.mm.1 NM_013524.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002413.mm.1 NM_013524.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002413.mm.1 TC0200000416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200002416.mm.1 NM_001177366.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002416.mm.1 NM_001177366.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002416.mm.1 NM_001177367.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002416.mm.1 NM_001177367.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002416.mm.1 NM_001289453.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002416.mm.1 NM_001289454.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002416.mm.1 NM_001289455.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002416.mm.1 NM_001289456.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002416.mm.1 NM_013524.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002416.mm.1 NM_013524.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002416.mm.1 TC0200000416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200002415.mm.1 NM_001177366.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002415.mm.1 NM_001177366.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002415.mm.1 NM_001177367.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002415.mm.1 NM_001177367.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002415.mm.1 NM_001289453.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002415.mm.1 NM_001289454.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002415.mm.1 NM_001289455.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002415.mm.1 NM_013524.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002415.mm.1 NM_013524.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002415.mm.1 TC0200000416.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 408566 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline