Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04242851_g1
          See other GM6760 GE Assays ›
          Gene Symbol:
          Gm6760
          Gene Name:
          predicted gene 6760
          Gene Aliases:
          EG627470
          Chromosome Location:
          Chr.X: 64151405 - 64152198 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04242851_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm6760
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177377.1 NP_001170848.1 1 - 2 253 75
          GenBank mRNA BC048649.1 - 1 - 2 148 75
          BU939216.1 - 1 - 2 254 75
          CN841342.1 - 1 - 2 164 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002331.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P136193 NM_001177377.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P136193 CN841342.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136193 NM_001177377.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136193 CN841342.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14641677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15102436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15209592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14711682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15142394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14782830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14741233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14641671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32426984 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00014631.mm.1 NM_001177377.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014631.mm.1 NM_001177377.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014631.mm.1 TC0X00002331.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00014632.mm.1 NM_001177377.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014632.mm.1 NM_001177377.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014632.mm.1 TC0X00002331.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline