Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04243026_g1
          See other GM7325 GE Assays ›
          Gene Symbol:
          Gm7325
          Gene Name:
          predicted gene 7325
          Gene Aliases:
          EG653016, Esgp
          Chromosome Location:
          Chr.17: 45600961 - 45602204 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04243026_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm7325
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177470.1 NP_001170941.1 1 - 2 133 96
          GenBank mRNA CJ062905.1 - 1 - 2 123 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002153.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114804 NM_001177470.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114804 NM_001177469.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114804 NM_001177468.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P31202116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31202129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31202126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31202111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13185818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31202110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31202118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31202120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31202121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13555867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31202133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13558994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13430561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13526744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31202130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31202127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13183932 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10451143 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451144 - -
          Affymetrix HT MG-430 PM Array Plate 1456242_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10451142 - -
          Mouse Genome 430 2.0 Array 1456242_at - -
          Mouse Genome 430 2.0 Array 1456242_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 653016_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 653016_at - -
          Affymetrix Mouse Clariom S Array PSR1700020181.mm.1 NM_001177468.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020181.mm.1 NM_001177468.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020181.mm.1 NM_001177469.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020181.mm.1 NM_001177469.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020181.mm.1 NM_001177470.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020181.mm.1 NM_001177470.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020181.mm.1 TC1700002153.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020182.mm.1 NM_001177470.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020182.mm.1 NM_001177470.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020182.mm.1 TC1700002153.mm.1 MTA TC
          Mouse Expression Set 430 1456242_at - -
          Mouse Expression Set 430 1456242_PM_at - -
          Murine Genome U74v2 Set 167088_r_at - -
          Murine Genome U74v2 Set 167088_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1456242_at - -
          Affymetrix HT Mouse Genome 430B Array 1456242_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline