Search
Search

4930480E11Rik| Gene Symbol: 4930480E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177966.2 | NP_001171437.1 | 1 | 1054 | 101 | |
| GenBank mRNA | AK015598.1 | - | 1 | 1055 | 101 | |
| BC116859.1 | - | 1 | 1054 | 40126178 | 101 | |
| BC116861.1 | - | 1 | 1054 | 40126180 | 101 | |
Set Membership: |
|