| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00002744.mm.1 |
NM_001195271.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002744.mm.1 |
NM_001195271.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002744.mm.1 |
TC0X00000408.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002743.mm.1 |
NM_001195271.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002743.mm.1 |
NM_001195271.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002743.mm.1 |
TC0X00000408.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002745.mm.1 |
NM_001195271.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002745.mm.1 |
NM_001195271.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002745.mm.1 |
TC0X00000408.mm.1 |
MTA TC |