Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04243952_s1
          See other OLFR122 GE Assays ›
          Gene Symbol:
          Olfr122
          Gene Name:
          olfactory receptor 122
          Gene Aliases:
          MOR263-10
          Chromosome Location:
          Chr.17: 37768642 - 37772620 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04243952_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr122
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146288.3 NP_666400.3 2 848 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr122

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2097814 NM_146288.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2097814 NM_146288.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P511669 NM_146288.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169881 - -
          Whole Mouse Genome G4122A A_51_P511669 NM_146288.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13325099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13350183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13182083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13593302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13579894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13325095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169859 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445144 - -
          Affymetrix Mouse Gene 1.1 ST Array 10445143 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3130315 - -
          Illumina mouse-6 v1.0 expression beadchip 3130315 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003130315 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2753678 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2753678 - -
          mouse-6 v1.1 (Illumina) scl8514.1.1_195-S - -
          Illumina Sentrix mouse-6.v1 bead array 3130315 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8514.1.1_195-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3130315 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2753678 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2753678 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0