Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04243954_s1
          See other 6720489N17RIK GE Assays ›
          Gene Symbol:
          6720489N17Rik
          Gene Name:
          RIKEN cDNA 6720489N17 gene
          Gene Aliases:
          9330128I06
          Chromosome Location:
          Chr.13: 62603015 - 62624182 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04243954_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 6720489N17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173381.1 NP_775557.1 3 2147 83
          GenBank mRNA AK033958.1 - 3 2131 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2184737 AK033958.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2184737 NM_173381.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161232 AK033958.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161232 NM_173381.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161232 AK033001.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112251 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10409991 - -
          Affymetrix HT MG-430 PM Array Plate 1457278_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440869_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10409990 - -
          Mouse Genome 430 2.0 Array 1457278_PM_at - -
          Mouse Genome 430 2.0 Array 1440869_PM_x_at - -
          Mouse Genome 430 2.0 Array 1440869_x_at - -
          Mouse Genome 430 2.0 Array 1457278_at - -
          Affymetrix Mouse Clariom S Array PSR1300013924.mm.1 NM_173381.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013924.mm.1 NM_173381.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013924.mm.1 TC1300002169.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013927.mm.1 NM_173381.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013927.mm.1 NM_173381.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013927.mm.1 TC1300002169.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013926.mm.1 NM_173381.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013926.mm.1 NM_173381.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013926.mm.1 TC1300002169.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013925.mm.1 NM_173381.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013925.mm.1 NM_173381.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013925.mm.1 TC1300002169.mm.1 MTA TC
          Mouse Expression Set 430 1440869_PM_x_at - -
          Mouse Expression Set 430 1457278_at - -
          Mouse Expression Set 430 1457278_PM_at - -
          Mouse Expression Set 430 1440869_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1457278_at - -
          Affymetrix HT Mouse Genome 430B Array 1440869_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1440869_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1457278_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259440 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline