Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04336752_s1
          See other IGIP GE Assays ›
          Gene Symbol:
          Igip
          Gene Name:
          IgA inducing protein
          Gene Aliases:
          6330403M23Rik
          Chromosome Location:
          Chr.18: 36300101 - 36301478 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04336752_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001267796.1 NP_001254725.1 1 1225 70
          GenBank mRNA AK078053.1 - 1 1225 70

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2420873 AI836787.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2420873 AK078053.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 953020 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2420873 AI836787.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2420873 AK078053.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14010251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13815607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13839663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14010291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623148 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1455538_PM_at - -
          Mouse Genome 430 2.0 Array 1455538_at - -
          Mouse Genome 430 2.0 Array 1455538_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800002433.mm.1 NM_001267796.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002433.mm.1 TC1800000324.mm.1 MTA TC
          Mouse Expression Set 430 1455538_PM_at - -
          Mouse Expression Set 430 1455538_at - -
          Affymetrix HT Mouse Genome 430B Array 1455538_at - -
          Affymetrix HT Mouse Genome 430B Array 1455538_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 364159 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline