Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301340.1 | NP_001288269.1 | 16 | 2122 | 72 | |
NM_001301341.1 | NP_001288270.1 | 18 | 2249 | 72 | ||
NM_001301343.1 | NP_001288272.1 | 16 | 2108 | 72 | ||
NM_001301344.1 | NP_001288273.1 | 15 | 2050 | 72 | ||
NM_001301345.1 | NP_001288274.1 | 15 | 1990 | 72 | ||
NM_025279.3 | NP_079555.1 | 16 | 2062 | 72 | ||
GenBank mRNA | AK088462.1 | - | 15 | 1976 | 72 | |
AK128891.1 | - | 16 | 2059 | 72 | ||
AK135519.1 | - | 16 | 2035 | 72 | ||
AK150848.1 | - | 16 | 2102 | 72 | ||
AK160887.1 | - | 16 | 2063 | 72 | ||
AK166656.1 | - | 16 | 2126 | 72 | ||
AK166927.1 | - | 16 | 2080 | 71 | ||
AK167600.1 | - | 16 | 2052 | 72 | ||
AK168918.1 | - | 12 | 1963 | 72 | ||
BC089328.1 | - | 16 | 2046 | 6402614 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon |